Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimethyl pyridine-2,6-dicarboxylate | 5453-67-8 | 195.17 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW007966100G

Encompass_Preferred

Dimethyl pyridine-2,6-dicarboxylate is a biochemical reagent primarily used as a biological material or organic compound for life science-related research. It presents as a white to off-white solid, stable under recommended storage conditions. This product is intended for research use only.

  • Used in laboratory chemicals and substance manufacturing
  • Solid appearance
  • Melting point 121-125°C
  • Boiling point 321.7°C at 760 mmHg

Catalog No. 50-004-47870


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.