Learn More
Medchemexpress LLC Dimethyl pyridine-2,6-dicarboxylate | 5453-67-8 | 195.17 | 250 G
SDP
Catalog No. 5000450040
Encompass_Preferred
Dimethyl pyridine-2,6-dicarboxylate is a biochemical reagent used as a biological material or organic compound for life science research. It is suitable for laboratory applications and appears as a white to off-white solid. It is for research use only.
  • Biochemical reagent for life science research
  • White to off-white solid appearance
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Keep container tightly sealed in a cool, well-ventilated area
Catalog No. 50-004-50040 Supplier Medchemexpress LLC Supplier No. 250GHYW007966
Dimethyl pyridine-2,6-dicarboxylate is a biochemical reagent used as a biological material or organic compound for life science research. It is suitable for laboratory applications and appears as a white to off-white solid. It is for research use only.
  • Biochemical reagent for life science research
  • White to off-white solid appearance
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Keep container tightly sealed in a cool, well-ventilated area