Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl (S)-(-)-Malate

Catalog No. 3743185G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-431-85G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 37-431-85G Supplier Sigma Aldrich Supplier No. 3743185G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This chiral synthon has been used to prepare cytochrome P450 metabolites of arachidonic acid, and cyclic sulfolanes with HIV-1 protease inhibition potential.

Specifications

CAS 13139-16-7
Melting Point 66°C to 69°C
Molecular Formula C11H21NO4
Linear Formula CH3CH2CH(CH3)CH(COOH)NHCOOC(CH3)3
Refractive Index n20/D 1.435
Quantity 25 g
MDL Number MFCD00038324
Synonym Dimethyl (S)-2-hydroxysuccinate; Dimethyl L-malate
Molecular Weight (g/mol) 231.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.