Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl (S)-(-)-Methylsuccinate 98.0+%, TCI America™
SDP

Catalog No. M12325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12325G Supplier TCI America Supplier No. M12325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63163-08-6
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00134572
InChI Key NFOQJNGQQXICBY-UHFFFAOYNA-N
Synonym Dimethyl (S)-(-)-Pyrotartarate, (S)-(-)-Methylsuccinic Acid Dimethyl Ester, (S)-(-)-Pyrotartaric Acid Dimethyl Ester
PubChem CID 9964174
IUPAC Name 1,4-dimethyl 2-methylbutanedioate
SMILES COC(=O)CC(C)C(=O)OC

Specifications

Color Colorless
Boiling Point 196°C
Quantity 5 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl (S)-(-)-Methylsuccinate
EINECSNumber (2)-0904
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.