Learn More
Medchemexpress LLC Dimethyl silicone oil | 63148-62-9 | MFCD00084411; MFCD00132673 | 1,125 g/mol (number-average) | [Si(CH3)2O]n | 25 ML
SDP
Catalog No. 5000243357
Encompass_Preferred
Dimethyl silicone oil is a low-viscosity polydimethylsiloxane (PDMS) fluid with a kinematic viscosity of 10 cSt at 25 °C (CAS 63148-62-9). It is a colorless to light yellow, hydrophobic, and chemically inert fluid commonly used in laboratory and industrial research as a lubricant, release agent, and solvent modifier.
  • Low kinematic viscosity of 10 cSt at 25 °C for easy flow
  • Hydrophobic and chemically inert, providing chemical compatibility
  • Low volatility and vapor pressure to reduce losses during use
  • Relative density ~0.93 g/cm³ for formulation and dosing calculations
  • Stable under recommended storage conditions for extended shelf life
  • Suitable as a lubricant, release agent, or solvent modifier in research applications
Catalog No. 50-002-43357 Supplier Medchemexpress LLC Supplier No. HY104035H25ML
Dimethyl silicone oil is a low-viscosity polydimethylsiloxane (PDMS) fluid with a kinematic viscosity of 10 cSt at 25 °C (CAS 63148-62-9). It is a colorless to light yellow, hydrophobic, and chemically inert fluid commonly used in laboratory and industrial research as a lubricant, release agent, and solvent modifier.
  • Low kinematic viscosity of 10 cSt at 25 °C for easy flow
  • Hydrophobic and chemically inert, providing chemical compatibility
  • Low volatility and vapor pressure to reduce losses during use
  • Relative density ~0.93 g/cm³ for formulation and dosing calculations
  • Stable under recommended storage conditions for extended shelf life
  • Suitable as a lubricant, release agent, or solvent modifier in research applications