Learn More
Medchemexpress LLC Dimethyl silicone oil | 63148-62-9 | [Si(CH3)2O]n | 50 ML
SDP
Catalog No. 5000242500
Encompass_Preferred
Dimethyl silicone oil is a polydimethylsiloxane fluid with a kinematic viscosity of 100 cSt at 25 °C. It is a colorless to light-yellow, chemically inert oil used in laboratory applications as a reagent, viscosity standard, lubricant, and formulation component.
  • Provides consistent 100 cSt viscosity at 25 °C
  • Chemically inert and low volatility for stable performance
  • Good thermal and oxidative stability across common lab temperatures
  • Hydrophobic and lubricating properties suitable for coatings and seals
  • Clear liquid with typical density ~0.96 g/cm³ for formulation use
  • Available in small-volume packaging suitable for research labs
Catalog No. 50-002-42500 Supplier Medchemexpress LLC Supplier No. HY104035E50ML
Dimethyl silicone oil is a polydimethylsiloxane fluid with a kinematic viscosity of 100 cSt at 25 °C. It is a colorless to light-yellow, chemically inert oil used in laboratory applications as a reagent, viscosity standard, lubricant, and formulation component.
  • Provides consistent 100 cSt viscosity at 25 °C
  • Chemically inert and low volatility for stable performance
  • Good thermal and oxidative stability across common lab temperatures
  • Hydrophobic and lubricating properties suitable for coatings and seals
  • Clear liquid with typical density ~0.96 g/cm³ for formulation use
  • Available in small-volume packaging suitable for research labs