Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl Sulfate

Catalog No. 503119096 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
1 L
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-311-9096 5 mL
D18630918LK 18 L
50-311-9095 100 mL
50-311-9108 500 mL
50-311-9107 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-311-9096 Supplier Sigma Aldrich Supplier No. D1863095ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethyl Sulfate

Specifications

CAS 366-18-7
Melting Point 70°C to 73°C
Boiling Point 273°C
Molecular Formula C10H8N2
Refractive Index n20/D 1.386
Linear Formula C10H8N2
Quantity 5 mL
MDL Number MFCD00006212
Synonym Sulfuric acid dimethyl ester
Molecular Weight (g/mol) 156.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.