Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl Sulfide

Catalog No. 47157725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-157-725ML 25 mL
47-157-7250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-157-725ML Supplier Sigma Aldrich Supplier No. 47157725ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethyl Sulfide

Specifications

CAS 75-18-3
Melting Point -98°C
Density 0.846 g/mL at 25°C
Boiling Point 38°C
Molecular Formula C2H6S
Refractive Index n20/D 1.435
Linear Formula (CH3)2S
MDL Number MFCD00008562
Quantity 25 mL
Synonym DMS; Methyl sulfide
Molecular Weight (g/mol) 62.13 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.