Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfide, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018671 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8671 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8671 Supplier MilliporeSigma Supelco Supplier No. 76048100MG
Add to Cart
Add to Cart

Dimethyl sulfide is found to be the most abundant volatile thioether, which can find applications as a flavor enhancing agent in food. It can undergo oxidation in the process of malting and kilning, to yield dimethyl sulfoxide (DMSO).

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Ignition Point 402°F
CAS 75-18-3
Vapor Pressure 26.24 psi (55°C)
Vapor Density 2.1 (vs. air)
Density 0.846 g/mL (at 25°C)
Flash Point -36°C
Physical Form Neat
Linear Formula (CH3)2 S
MDL Number MFCD00008562
Melting Point -98°C
Boiling Point 38°C
Synonym DMS; Methyl sulfide
Solubility Soluble in water
Type Analytical Standard
Quantity 100 mg
Formula Weight 62.13
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.