Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-pentanol, 99+%

Catalog No. AC149380025
Change view
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149380025 2.5 L Glass bottle
AC149380050 5 mL Glass bottle
AC149385000 500 mL Glass bottle
AC149380010 1 L Glass bottle
AC149380100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. AC149380025 Supplier Thermo Scientific Chemicals Supplier No. 149380025
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 108-11-2 | C6H14O | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-11-2
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004550
InChI Key WVYWICLMDOOCFB-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanol, 2-pentanol, 4-methyl, isobutylmethylcarbinol, 2-methyl-4-pentanol, methyl amyl alcohol, methyl isobutyl carbinol, mibc, 1,3-dimethylbutanol, 4-methylpentanol-2, isobutylmethylmethanol
PubChem CID 7910
IUPAC Name 4-methylpentan-2-ol
SMILES CC(C)CC(C)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -90.0°C
Color Colorless
Density 0.8000g/mL
Boiling Point 132.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4100 to 1.4120
Linear Formula (CH3)2CHCH2CH(OH)CH3
Quantity 2.5 L
Beilstein 01, 410
Specific Gravity 0.8
Solubility Information Solubility in water: 20g/L. Other solubilities: misscible with most common organic solvents
Formula Weight 102.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-551-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.