Learn More
Medchemexpress LLC Dimethyl sulfone | 67-71-0 | MFCD00007566 | 99.6% | 94.13 g/mol | C2H6O2S | 5g
SDP
Catalog No. 5000267895
Encompass_Preferred
Dimethyl sulfone (Methyl Sulfonyl Methane) is a metabolic product of endogenous methanethiol metabolism and intestinal bacterial metabolism Dimethyl sulfone inhibits choriocapillary endothelial (CCE) cell proliferation also has many biological effects including antiinflammatory antioxidant and local anesthetic effects that could be neuroprotective[1][2][3]
Catalog No. 50-002-67895 Supplier Medchemexpress LLC Supplier No. HYY13145G
Dimethyl sulfone (Methyl Sulfonyl Methane) is a metabolic product of endogenous methanethiol metabolism and intestinal bacterial metabolism Dimethyl sulfone inhibits choriocapillary endothelial (CCE) cell proliferation also has many biological effects including antiinflammatory antioxidant and local anesthetic effects that could be neuroprotective[1][2][3]