Learn More
Medchemexpress LLC Dimethyl sulfone | 67-71-0 | MFCD00007566 | 99.6% | 94.133 g/mol | C2H6O2S | 500g
SDP
Catalog No. 5000267808
Encompass_Preferred
Dimethyl sulfone (Methyl Sulfonyl Methane) is a metabolic product of endogenous methanethiol metabolism and intestinal bacterial metabolism Dimethyl sulfone inhibits choriocapillary endothelial (CCE) cell proliferation also has many biological effects including antiinflammatory antioxidant and local anesthetic effects that could be neuroprotective[1][2][3]
Catalog No. 50-002-67808 Supplier Medchemexpress LLC Supplier No. HYY1314500G
Dimethyl sulfone (Methyl Sulfonyl Methane) is a metabolic product of endogenous methanethiol metabolism and intestinal bacterial metabolism Dimethyl sulfone inhibits choriocapillary endothelial (CCE) cell proliferation also has many biological effects including antiinflammatory antioxidant and local anesthetic effects that could be neuroprotective[1][2][3]