Learn More
Medchemexpress LLC HY-Y0320 100mL , Dimethyl sulfoxide CAS:67-68-5 Purity:>98%
SDP
Catalog No. 502259283
Encompass_Preferred
HY-Y0320 100mL Dimethyl sulfoxide CAS: 67-68-5 Dimethyl sulfoxide (DMSO) is an aprotic solvent that dissolves both polar and nonpolar compounds. Dimethyl sulfoxide has anti-freezing and bacteriostatic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9283 Supplier Medchemexpress LLC Supplier No. HYY0320100ML
HY-Y0320 100mL Dimethyl sulfoxide CAS: 67-68-5 Dimethyl sulfoxide (DMSO) is an aprotic solvent that dissolves both polar and nonpolar compounds. Dimethyl sulfoxide has anti-freezing and bacteriostatic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.