Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide, 67-68-5, MFCD00002089, 1L
Catalog No. NC0328323
Encompass
Molecular Formula: (CH3)2SO, Molecular Weight: 78.13, Beilstein Registry Number: 506008, Assay: ≥99.5% (GC)
Catalog No. NC0328323 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D45401L
Molecular Formula: (CH3)2SO, Molecular Weight: 78.13, Beilstein Registry Number: 506008, Assay: ≥99.5% (GC)