Learn More
Crescent Chemical Co Inc DIMETHYL SULFOXIDE
SDP
Supplier:  Crescent Chemical Co Inc SE20385.02
Encompass_Preferred
Dimethyl sulfoxide research grade Highly active solvent and pharmaceutical vehicle; for freezing cells. For gradient centrifugation (1). Determination of cysteine and cystine in proteins (2). Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Num
Catalog No. 50-247-521