Learn More
Crescent Chemical Co Inc DIMETHYL SULFOXIDE
SDP
Catalog No. 50247520
Encompass_Preferred
Dimethyl sulfoxide research grade Highly active solvent and pharmaceutical vehicle; for freezing cells. For gradient centrifugation (1). Determination of cysteine and cystine in proteins (2). Size - 250ML Storage Conditions - +15 °C TO +30 °C Catalog
Catalog No. 50-247-520 Supplier Crescent Chemical Co Inc Supplier No. SE20385.01
Dimethyl sulfoxide research grade Highly active solvent and pharmaceutical vehicle; for freezing cells. For gradient centrifugation (1). Determination of cysteine and cystine in proteins (2). Size - 250ML Storage Conditions - +15 °C TO +30 °C Catalog