Learn More
U.S. Pharmacopeia Dimethyl Sulfoxide, 67-68-5, MFCD00002089, 3g
Catalog No. NC0314930
Encompass
Molecular formula C2H6OS, Molecular weight 78.13, Melting Point 64.4 - 66.2 °F (18 - 19 °C), Boiling Point 372.2 °F (189 °C), Synonyms: DMSO, Methyl sulfoxide
Catalog No. NC0314930 Supplier U.S. Pharmacopeia Supplier No. 1211006
Molecular formula C2H6OS, Molecular weight 78.13, Melting Point 64.4 - 66.2 °F (18 - 19 °C), Boiling Point 372.2 °F (189 °C), Synonyms: DMSO, Methyl sulfoxide