Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jade Scientific, Inc DIMETHYL SULFOXIDE 4L
SDP

Catalog No. 50326136 Shop All Jade Scientific, Inc Products
Encompass_Preferred

Dimethyl Sulfoxide 4l. 67-68-5 MFCD00002089

Catalog No. 50-326-136 Supplier Jade Scientific, Inc Supplier No. BJBB0824
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl Sulfoxide 4l. 67-68-5 MFCD00002089

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.