Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide | 67-68-5 | MFCD00002089 | 500 mL
Catalog No. NC0592255
Encompass
Dimethyl sulfoxide | Purity: ≥99.7% | Mol Wt: 78.13 | 67-68-5 | MFCD00002089 | 500 mL
Catalog No. NC0592255 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 34869500ML
Dimethyl sulfoxide | Purity: ≥99.7% | Mol Wt: 78.13 | 67-68-5 | MFCD00002089 | 500 mL