Learn More
Cambridge Isotope Laboratories Dimethyl Sulfoxide-d6 (d, 99.9%) +1% V/v Tms-50 Grams
Catalog No. NC9954226
Encompass_Preferred
Dimethyl Sulfoxide-d6 (d, 99.9%) +1% V/v Tms-50 Grams
Catalog No. NC9954226 Supplier Cambridge Isotope Laboratories Supplier No. DLM10TA50
Dimethyl Sulfoxide-d6 (d, 99.9%) +1% V/v Tms-50 Grams