Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jade Scientific, Inc DIMETHYL SULFOXIDE 56L
SDP

Supplier:  Jade Scientific, Inc BJCS08156

Encompass_Preferred

Dimethyl Sulfoxide 56l. 67-68-5 MFCD00002089

Catalog No. 50-326-227


May include imposed supplier surcharges.
Add to Cart
Request a Quote
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.