Learn More
Medchemexpress LLC Dimethyl sulfoxide | 67-68-5 | MFCD00002089 | 100.0% | 78.13 | C2H6OS | 1mL
SDP
Catalog No. 5000268703
Encompass_Preferred
Dimethyl sulfoxide (Standard) is the analytical standard of Dimethyl sulfoxide This product is intended for research and analytical applications Dimethyl sulfoxide (DMSO) is an aprotic solvent that dissolves both polar and nonpolar compounds Dimethyl sulfoxide has anti-freezing and bacteriostatic properties[1][2]
Catalog No. 50-002-68703 Supplier Medchemexpress LLC Supplier No. HYY0320R1ML
Dimethyl sulfoxide (Standard) is the analytical standard of Dimethyl sulfoxide This product is intended for research and analytical applications Dimethyl sulfoxide (DMSO) is an aprotic solvent that dissolves both polar and nonpolar compounds Dimethyl sulfoxide has anti-freezing and bacteriostatic properties[1][2]