Learn More
Medchemexpress LLC Dimethyl sulfoxide | 67-68-5 | MFCD00002089 | 100.0% | 78.13 g/mol | C2H6OS | 100mL
SDP
Catalog No. 5000265891
Encompass_Preferred
Dimethyl sulfoxide (DMSO) is an aprotic solvent that can dissolve water-insoluble therapeutic and toxic agents Dimethyl sulfoxide (DMSO) has a strong affinity for water and has the ability to rapidly penetrate or enhance the penetration of other substances through biological membranes Dimethyl sulfoxide also has potential free radical scavenging and anticholinesterase effects and may affect coagulation activity Dimethyl sulfoxide also induces histamine release from mast cells but is thought to have low systemic toxicity[1] br/ Low endotoxin can be used in various biochemical experiments such as drug dissolution
Catalog No. 50-002-65891 Supplier Medchemexpress LLC Supplier No. HYY0320C100ML
Dimethyl sulfoxide (DMSO) is an aprotic solvent that can dissolve water-insoluble therapeutic and toxic agents Dimethyl sulfoxide (DMSO) has a strong affinity for water and has the ability to rapidly penetrate or enhance the penetration of other substances through biological membranes Dimethyl sulfoxide also has potential free radical scavenging and anticholinesterase effects and may affect coagulation activity Dimethyl sulfoxide also induces histamine release from mast cells but is thought to have low systemic toxicity[1] br/ Low endotoxin can be used in various biochemical experiments such as drug dissolution