Learn More
Medchemexpress LLC Dimethyl sulfoxide | 67-68-5 | MFCD00002089 | 100.0% | 78.13 g/mol | C2H6OS | 10mL
SDP
Catalog No. 5000269337
Encompass_Preferred
Dimethyl sulfoxide (DMSO) meets analytical specification of Ch P is an aprotic solvent that dissolves polar and non-polar compounds including water-insoluble therapeutic and toxic agents Dimethyl sulfoxide (DMSO) has a strong affinity for water and can rapidly penetrate or enhance the penetration of other substances into biological membranes Dimethyl sulfoxide also has potential free radical scavenging and anticholinesterase effects and may affect coagulation activity Dimethyl sulfoxide also induces histamine release from mast cells but is thought to have low systemic toxicity Dimethyl sulfoxide also exhibits antifreeze and antibacterial properties[1][2][3] br/ MCE provides Dimethyl sulfoxide that complies with the inspection standards (Ch P) of Part 4 of the Chinese Pharmacopoeia (2020 Edition) Amicrobic low endotoxin can be used in various biochemical experiments such as drug dissolution
Catalog No. 50-002-69337 Supplier Medchemexpress LLC Supplier No. HYY032010ML
Dimethyl sulfoxide (DMSO) meets analytical specification of Ch P is an aprotic solvent that dissolves polar and non-polar compounds including water-insoluble therapeutic and toxic agents Dimethyl sulfoxide (DMSO) has a strong affinity for water and can rapidly penetrate or enhance the penetration of other substances into biological membranes Dimethyl sulfoxide also has potential free radical scavenging and anticholinesterase effects and may affect coagulation activity Dimethyl sulfoxide also induces histamine release from mast cells but is thought to have low systemic toxicity Dimethyl sulfoxide also exhibits antifreeze and antibacterial properties[1][2][3] br/ MCE provides Dimethyl sulfoxide that complies with the inspection standards (Ch P) of Part 4 of the Chinese Pharmacopoeia (2020 Edition) Amicrobic low endotoxin can be used in various biochemical experiments such as drug dissolution