Learn More
Medchemexpress LLC Dimethyl sulfoxide | 67-68-5 | MFCD00002089 | 99.9% | 78.13 g/mol | C2H6OS | 5mL
SDP
Catalog No. 5000270020
Encompass_Preferred
Dimethyl sulfoxide cryoprotectant vitrification agent (DMSO cryoprotectant vitrification agent) is a biochemical reagent that can be used as a biological material or organic compound for life science related research[1] This product has been sterilized and has no endotoxin It can be used for long-term cell cryopreservation and will not affect the normal growth of cells
Catalog No. 50-002-70020 Supplier Medchemexpress LLC Supplier No. HYY0320D5ML
Dimethyl sulfoxide cryoprotectant vitrification agent (DMSO cryoprotectant vitrification agent) is a biochemical reagent that can be used as a biological material or organic compound for life science related research[1] This product has been sterilized and has no endotoxin It can be used for long-term cell cryopreservation and will not affect the normal growth of cells