Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, ≥99.5%, Reagent Grade, Solstice

Catalog No. 60047972 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
1 gal.
5 L
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-972 5 L Poly Bottle
60-047-963 100 mL Poly Bottle
60-047-971 500 mL Poly Bottle
60-047-966 1 L Poly Bottle
60-047-967 2.5 L Poly Bottle
60-047-965 1 gal. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-047-972 Supplier Solstice Supplier No. D58795L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.5°C
Color Colorless
Density 1.10g/mL
Quantity 5 L
Explosion Limit 42% (63°F)
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Odor Characteristic
Packaging Poly Bottle
Physical Form Liquid
Vapor Pressure 0.42mmHg (20°C)
Linear Formula (CH3)2SO
Beilstein 506008
Boiling Point 189°C
Formula Weight 78.13g/mol
Percent Purity ≥99.5%
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
RTECSNumber 200-664-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.