Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, ≥99%, MP Biomedicals™

Catalog No. ICN19605525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
0.5 L
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19605525 25 mL
ICN19605580 100 mL
ICN19605590 0.5 L
ICN19605591 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19605525 Supplier MP Biomedicals, Inc Supplier No. 0219605525
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Quantity 25 mL
Physical Form Liquid
Assay Percent Range ≥99%
IUPAC Name methylsulfinylmethane
Formula Weight 78.1
Percent Purity ≥99%
Chemical Name or Material Dimethyl sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.