Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™

Catalog No. ICN19018690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19018690 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19018690 Supplier MP Biomedicals, Inc Supplier No. 0219018690
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimethyl Sulfoxide is used to enhance dermal absorption of many chemicals.

  • Dimethyl sulfoxide is used to enhance dermal absorption of many chemicals.
  • A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers.
  • Used in antifreeze or hydraulic fluids and as a cryopreservative for cell cultures.
  • Used in the oxidation of thiols and disulfides to sulfonic acids.
  • Used as a PCR cosolvent to help improve yields, especially in long PCR.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Density 1.104 g/cm3 at 20 °C
Quantity 500 mL
Assay Percent Range 99+%
Formula Weight 78.13
Chemical Name or Material Dimethyl Sulfoxide
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.