Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, ACS, 99.9% min

Catalog No. AA36480M6
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 x 4 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36480M6 4 x 4 L
AA36480AP 500 mL
AA36480K2 1 L
AA36480M1 4 x 1 L
AA36480K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AA36480M6 Supplier Thermo Scientific Chemicals Supplier No. 036480M6
Only null left
Add to Cart

CAS: 67-68-5 | C2H6OS | 78.13 g/mol

It is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Density 1.101 g/mL
Quantity 4 x 4 L
Flash Point 87°C (189°F)
Physical Form Clear,colorless Liquid
Refractive Index 1.479
Assay Percent Range 99.9%
Beilstein 506008
Boiling Point 189°C
Solubility Information Miscible with water,diethyl ether,ethanol,acetone,ether,carbon tetrachloride and ethyl acetate.
Formula Weight 78.13
Grade ACS
Chemical Name or Material Dimethyl sulfoxide, For analysis ACS
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P370+P378q-P501c
EINECSNumber 200-664-3
RTECSNumber PV6210000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.