Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, ACS, 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18610226 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
200 L
Packaging:
Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-226 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-610-226 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1287200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-68-5
Molecular Weight (g/mol) 78.13

Specifications

CAS Max % 1
Quantity 200 L
Packaging Drum
Residue after Evaporation 0.0001
Titratable Acid 0.001 meq/g
Water 0.001
Boiling Point 189°C
Percent Purity 0.999
Grade Reagent
Chemical Name or Material Dimethyl Sulfoxide

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.