Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, Analytical Standard, ≥99.95% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020405 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0405 1 mL
11-101-8317 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0405 Supplier MilliporeSigma Supelco Supplier No. 945631MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For information on dimethyl sulfoxide miscibility, please visit the following link:Dimethyl Sulfoxide Miscibility/Immiscibility Table

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 16°C to 19°C
CAS 67-68-5
Density 1.10 g/mL
Quantity 1 mL
Physical Form Neat
Vapor Pressure 0.42 mmHg (20°C)
Linear Formula (CH3)2 SO
MDL Number MFCD00002089
Synonym DMSO
Boiling Point 189°C
Solubility Information H2O: miscible (completely)
Formula Weight 78.13
Percent Purity ≥99.95% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.