Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, for HPLC, GC and spectrophotometry, >99.5%, Solstice

Catalog No. 6001860 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-018-60 4 x 4 L Glass Bottle
60-018-59 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-018-60 Supplier Solstice Supplier No. 0814

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Absorbance 0.5 AU max. at 275 nm (UV), 0.2 AU max. at 300 nm (UV), 0.02 AU max. at 350 nm (UV), 0.005 AU max. at 400 nm (UV)
CAS Max % 100.0000%
Density 1.0954 g/cm3
Quantity 4 x 4 L
Flash Point 88°C
Packaging Glass Bottle
Water 0.04% max.
Beilstein 506008
Boiling Point 189°C
Solubility Information Miscible in Water at 20°C
Formula Weight 78.13
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material Dimethyl Sulfoxide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.