Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, Bioreagent

Catalog No. AAJ66650AE
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66650AE 100 mL
AAJ66650AD 50 mL
AAJ66650AK 250 mL
AAJ66650AP 500 mL
AAJ66650K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AAJ66650AE Supplier Thermo Scientific Chemicals Supplier No. J66650AE
Only null left
Add to Cart

CAS: 67-68-5 | C2H6OS | 78.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum.

Solubility
Soluble in water, methanol, acetone, ether, benzene, chloroform.

Notes
Submicron filtered. Store at Room Temperature (15-30°C), desiccate. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Color Colorless
Density 1.101 g/mL
Quantity 100 mL
Flash Point 87°C (189°F)
Odor Mild
Physical Form Liquid
Refractive Index 1.479
Beilstein 506008
Boiling Point 189°C
Solubility Information Soluble in water,methanol,acetone,ether,benzene,chloroform.
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Chemical Name or Material Dimethyl sulfoxide, Bioreagent
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P370+P378q-P501c
EINECSNumber 200-664-3
RTECSNumber PV6210000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.