Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide (Certified ACS), Fisher Chemical™

Catalog No. p-215904 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
HDPE Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D128-1 1 L HDPE Bottle
D128-4 4 L HDPE Bottle
D128-500 500 mL HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D128-1
Only null left
Add to Cart

Note: Sold for nonmedical use only. Not to be used or sold as a drug for humans or animals

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Acidity Pass Test (USP)
Appearance Clear, Colorless Liquid
Decomposition Information >190°C
Density 1.100g/cm³
Quantity 1 L
Flash Point 87°C (188.6°F)
Identification Pass Test (USP, EP/BP)
Packaging HDPE Bottle
Physical Form Liquid
Residue after Evaporation 0.01% max.
Assay Percent Range 99.9 % (CH3)2SO Minimum
Titratable Acid 0.001mEq/g max.
Vapor Pressure 0.55mbar at 20°C
Linear Formula (CH3)2SO
Viscosity 1.98 mPa/s at 25°C
Water 0.1% max.
Boiling Point 189°C
Solubility Information Soluble starch and sulfate: Pass Test (USP), 15 ppm max. (EP/BP)
Formula Weight 78.13
Percent Purity ≥99.9%
Grade ACS
Concentration or Composition (by Analyte or Components) Dextrin: Pass Test (USP, EP/BP)
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:2
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.