Learn More
Cambridge Isotope Laboratories Dimethyl Sulfoxide-d6, 2206-27-1, MFCD00002090, 100 g
Catalog No. NC9354739
Encompass_Preferred
Chemical Formula: CD3SOCD3, MW: 84.17, 99.9%, Synonyms: DMSO-D6; Methyl sulfoxide-D6; Hexadeuterodimethyl sulfoxide.
Catalog No. NC9354739 Supplier Cambridge Isotope Laboratories Supplier No. DLM10100
Chemical Formula: CD3SOCD3, MW: 84.17, 99.9%, Synonyms: DMSO-D6; Methyl sulfoxide-D6; Hexadeuterodimethyl sulfoxide.