Learn More
Cambridge Isotope Laboratories Dimethyl Sulfoxide-D6 (D, 99.9%), 2206-27-1, Mfcd00002090, 10X0.75 Ml
Catalog No. NC9800685
Encompass_Preferred
Chemical Formula: CD3SOCD3, MW: 84.17, 99.9%, Synonyms: DMSO-D6; Methyl sulfoxide-D6; Hexadeuterodimethyl sulfoxide.
Catalog No. NC9800685 Supplier Cambridge Isotope Laboratories Supplier No. DLM1010X0.75
Chemical Formula: CD3SOCD3, MW: 84.17, 99.9%, Synonyms: DMSO-D6; Methyl sulfoxide-D6; Hexadeuterodimethyl sulfoxide.