Learn More
Cambridge Isotope Laboratories Dimethyl Sulfoxide-d6 100% (d, 99.96%) +0.03% V/v Tms-10x0.75 Ml
Catalog No. NC9702325
Encompass_Preferred
Dimethyl Sulfoxide-d6 100% (d, 99.96%) +0.03% V/v Tms-10x0.75 Ml
Catalog No. NC9702325 Supplier Cambridge Isotope Laboratories Supplier No. DLM34TC10X0.75
Dimethyl Sulfoxide-d6 100% (d, 99.96%) +0.03% V/v Tms-10x0.75 Ml