Learn More
Cambridge Isotope Laboratories Dimethyl Sulfoxide-d6 100% (d, 99.96%), 2206-27-1, MFCD00002090, 10X0.5 mL
Catalog No. NC9031568
Encompass_Preferred
Chemical Formula: CD3SOCD3, MW: 84.17, 99.96%, Synonyms: DMSO-D6; Methyl sulfoxide-D6; Hexadeuterodimethyl sulfoxide.
Catalog No. NC9031568 Supplier Cambridge Isotope Laboratories Supplier No. DLM3410X0.5
Chemical Formula: CD3SOCD3, MW: 84.17, 99.96%, Synonyms: DMSO-D6; Methyl sulfoxide-D6; Hexadeuterodimethyl sulfoxide.