Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide-d6 34
Catalog No. 502897163
Encompass
Dimethyl sulfoxide-d6 (DMSO-d6) is a deuterated NMR solvent containing 0.03% (v/v) TMS (tetramethylsilane). It is useful in NMR-based research and analyses. It undergoes photodissociation to generate CD3 radical photoproducts which have been analyzed by infrared diode laser-absorption spectroscopy. Dissociation dynamics of DMSO-d6 at 193nm was examined using photo fragment translational spectroscopy method.
Catalog No. 50-289-7163 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 41793910X0.75ML
 Restricted Item
Add to Cart
Add to Cart
Dimethyl sulfoxide-d6 (DMSO-d6) is a deuterated NMR solvent containing 0.03% (v/v) TMS (tetramethylsilane). It is useful in NMR-based research and analyses. It undergoes photodissociation to generate CD3 radical photoproducts which have been analyzed by infrared diode laser-absorption spectroscopy. Dissociation dynamics of DMSO-d6 at 193nm was examined using photo fragment translational spectroscopy method.