Learn More
Cambridge Isotope Laboratories Dimethyl Sulfoxide-d6 (d, 99.9%) +0.05% V/v Tms, 2206-27-1, MFCD00002090, 50 g
Catalog No. NC9737783
Encompass_Preferred
Chemical Formula: CD3SOCD3, MW: 84.17, 99.9%, Synonyms: DMSO-D6; Methyl sulfoxide-D6; Hexadeuterodimethyl sulfoxide.
Catalog No. NC9737783 Supplier Cambridge Isotope Laboratories Supplier No. DLM10TB50
Add to Cart
Add to Cart
Chemical Formula: CD3SOCD3, MW: 84.17, 99.9%, Synonyms: DMSO-D6; Methyl sulfoxide-D6; Hexadeuterodimethyl sulfoxide.