Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide-d6, 99.9 Atom% D, Spectrum™ Chemical
SDP

Catalog No. 18602252 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
50 g
10 x 0.75 mL
10 x 1 g
Packaging:
Ampule
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-252 10 x 0.75 mL Ampule
18-602-246 10 x 1 g Ampule
18-602-247 5 g Ampule
18-602-248 10 g Ampule
18-602-250 50 g Ampule
18-602-249 25 g Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-252 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D2493135
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D2493, 2206-27-1, CD3SOCD3

Spectrum™ Chemical Dimethyl Sulfoxide-d6, 99.9 Atom Percent D - Spectrum offers a broad range of high quality deuterated products for all your routine synthesis and structural analysis needs and is the preferred brand for deuterated NMR solvents worldwide. We offer Isotopic Enrichment ranges from 97 atom % D for routine use to 100 atom % D for the most demanding applications.

Specifications

Chemical Name or Material Dimethyl Sulfoxide to d6, 99.9 Atom Percent D
Color Colorless
Packaging Ampule
Quantity 10 x 0.75 mL
CAS 2206-27-1
CAS Max % 100%
Molecular Weight (g/mol) 87.14
Percent Purity 99.9%
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.