Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, For inorganic trace analysis, 99.99995% (metals basis), MilliporeSigma™ Supelco™

Catalog No. 111018372 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8372 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8372 Supplier MilliporeSigma Supelco Supplier No. 019341L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For information on dimethyl sulfoxide miscibility, please visit the following link:Dimethyl Sulfoxide Miscibility/Immiscibility Table

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym DMSO
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 16°C to 19°C; 16°C to 19°C
Color Colorless
Density 1.10 g/mL
Boiling Point 189°C; 189°C
Refractive Index n20/D 1.479
Linear Formula (CH3)2 SO
Quantity 1 L
Vapor Density 2.7 (vs. air)
Vapor Pressure 0.42 mmHg (20°C)
Formula Weight 78.13
Percent Purity ≥99.9% (GC)
Grade For inorganic trace analysis
Show More Show Less
RTECSNumber PV6210000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.