Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide, Solstice™

Catalog No. 60090240 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-240 6 x 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-240 Supplier Solstice Supplier No. 348696X100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ Plus, for HPLC, ≥99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
PubChem CID 679
ChEBI CHEBI:28262

Specifications

Quantity 6 x 100 mL
Packaging Glass Bottle
Physical Form Liquid
Percent Purity min. 99.70 %
Grade HPLC
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.