Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide
Catalog No. NC1758165
Encompass
Dimethyl sulfoxide | Purity: 99.7% | Mol Wt: 78.13 | 67-68-5 | MFCD00002089 | 1L
Catalog No. NC1758165 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348691L
Dimethyl sulfoxide | Purity: 99.7% | Mol Wt: 78.13 | 67-68-5 | MFCD00002089 | 1L