Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide (HPLC Grade), Fisher Chemical

Catalog Number D1594
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methanesulfinylmethane
CS(C)=O
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
methanesulfinylmethane
CS(C)=O
Melting Point 18.4°C
Absorbance 0.01 AU max., 0.02 AU max. at 335nm, 0.02 AU max. at 330nm, 0.06 AU max. at 310nm, 0.1 AU max. at 300nm, 0.25 AU max. at 280nm, 0.2 AU max. at 290nm, 0.3 AU max. at 275nm, 0.4 AU max. at 270nm, 1.00 AU max. at 263nm
Color Colorless
Decomposition Information >190°C
Density 1.100g/cm³
Boiling Point 189°C
Quantity 4 L
Flash Point 87°C (188.6°F)
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Refractive Index 1.4770 to 1.4800 (at 25°C)
Residue after Evaporation 0.005% Max
Assay Percent Range 99.8% min by GC
Titratable Acid 0.001mEq/g max.
Linear Formula (CH3)2SO
Vapor Pressure 0.55mbar at 20°C
Viscosity 1.98 mPa/s at 25°C
Water 0.01% max.
Formula Weight 78.13
Percent Purity ≥99.9%
Grade Mobile phase for HPLC applications
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:2
Instability:0
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit