Learn More
Crescent Chemical Co Inc Dimethyl Sulfoxide, 67-68-5, MFCD00002089, 250mL
SDP
Supplier:  Crescent Chemical Co Inc SE39757.02
Encompass_Preferred
Molecular Formula C2H6OS, Molecular Weight 78.134, Molecular Biology Grade, DNase/RNase not detected. Storage Conditions: +15 °C TO +30 °C
Catalog No. 50-248-437
 Restricted Item