Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide puriss.
Catalog No. 502899276
Encompass
Dimethyl sulfoxide (DMSO) is a polar solvent. DMSO activated by electrophile (oxalyl chloride) has been used to oxidize various alcohols (primary secondary allylic benzylic hindered and bicyclic). Carbonyl compounds are formed as reaction products. It has been proposed as an alternate solvent for methyl cellosolve for use in ninhydrin reaction due to its high stability.
Catalog No. 50-289-9276 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 41640100ML
Dimethyl sulfoxide (DMSO) is a polar solvent. DMSO activated by electrophile (oxalyl chloride) has been used to oxidize various alcohols (primary secondary allylic benzylic hindered and bicyclic). Carbonyl compounds are formed as reaction products. It has been proposed as an alternate solvent for methyl cellosolve for use in ninhydrin reaction due to its high stability.