Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, Puriss p.a, ACS Reagent, ≥99.9% (GC), Solstice

Catalog No. 60046840 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-840 500 mL Poly Bottle
60-046-838 1 L Poly Bottle
60-046-837 100 mL Poly Bottle
60-046-839 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-840 Supplier Solstice Supplier No. 41640500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent,≥99.9% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Color Colorless
Density 1.10g/mL
Quantity 500 mL
Explosion Limit 42%
Ignition Point 573°C
Packaging Poly Bottle
Refractive Index 1.478 to 1.48
Linear Formula (CH3)2SO
Vapor Pressure 0.42mmHg (20°C)
Water 0 to 0.1% (Coulometric)
Beilstein 506008
IUPAC Name methanesulfinylmethane
Formula Weight 78.13g/mol
Percent Purity ≥99.9% (GC)
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
RTECSNumber 200-664-3
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.