Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide puriss500ML
Catalog No. 502899223
Encompass
Dimethyl sulfoxide (DMSO) is a polar solvent. DMSO activated by electrophile (oxalyl chloride) has been used to oxidize various alcohols (primary secondary allylic benzylic hindered and bicyclic). Carbonyl compounds are formed as reaction products. It has been proposed as an alternate solvent for methyl cellosolve for use in ninhydrin reaction due to its high stability.
Catalog No. 50-289-9223 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 41640500ML
Dimethyl sulfoxide (DMSO) is a polar solvent. DMSO activated by electrophile (oxalyl chloride) has been used to oxidize various alcohols (primary secondary allylic benzylic hindered and bicyclic). Carbonyl compounds are formed as reaction products. It has been proposed as an alternate solvent for methyl cellosolve for use in ninhydrin reaction due to its high stability.