Learn More
Sigma Aldrich Fine Chemicals Biosciences Dimethyl sulfoxide Reagent
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D5879100ML
Encompass
Dimethyl sulfoxide is an amphipathic molecule soluble in both aqueous and organic solvents. It is commonly used as an aprotic solvent and also as a reagent in organic synthesis.
Catalog No. 50-289-7417